missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FBXL4 (aa 26-110) Control Fragment Recombinant Protein

Product Code. 30209891
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209891

Brand: Invitrogen™ RP94678

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56072 (PA5-56072. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains at least 9 tandem leucine-rich repeats.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UKA2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26235
Name Human FBXL4 (aa 26-110) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI836810; FBL4; FBL5; F-box and leucine rich repeat protein 4; F-box and leucine-rich repeat protein 4; F-box protein FBL4/FBL5; F-box/LRR-repeat protein 4; Fbxl4; MTDPS13
Common Name FBXL4
Gene Symbol FBXL4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ATRGEMMNTHRAIESNSQTSPLNAEVVQYAKEVVDFSSHYGSENSMSYTMWNLAGVPNVFPSSGDFTQTAVFRTYGTWWDQCPSA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.