Learn More
Human Fatty Acid Synthase/FASN Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB59271-SP
Product Code. 18742218
  • £167.00 / 25µg
Description
Fatty Acid Synthase/FASN Monoclonal specifically detects Fatty Acid Synthase/FASN in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fatty Acid Synthase/FASN
Monoclonal
LYOPH
Western Blot 0.25 ug/mL
P49327
FASN
E. coli-derived recombinant human Fatty Acid Synthase/FASN Ser10-Cys212 Accession # P49327
25 μg
Primary
Detects Fatty Acid Synthase/FASN in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) Ephrin-A2 is observed and no cross-reactivity with recombinant human (rh) Ephrin-A3, -B3, rmEphrin-A4, -A5, -B1 or -B2 is observed.
Human
Purified
Western Blot
647114
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASN, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2194
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b