missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FATP4 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3650G-100UG
This item is not returnable.
View return policy
Description
FATP4/SLC27A4 Monoclonal specifically detects FATP4/SLC27A4 in Human samples. It is validated for Flow Cytometry.
Specifications
| FATP4/SLC27A4 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| ACSVL4, ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, SLC27A4, solute carrier family 27 (fatty acid transporter), member 4 | |
| NS0 mouse myeloma cell line transfected with human FATP4 Met1-Leu643 Accession # Q6P1M0 | |
| RUO | |
| 10999 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 342142 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6P1M0 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FATP4. Stains human FATP4-transfected cells but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction