missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FATP2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4659G-100UG
This item is not returnable.
View return policy
Description
FATP2/SLC27A2 Monoclonal specifically detects FATP2/SLC27A2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| FATP2/SLC27A2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| ACSVL1, ACSVL1FATP-2, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.3, EC 6.2.1.7, FACVL1, FACVL1very long-chain fatty-acid-coenzyme A ligase 1, FATP2THCA-CoA ligase, Fatty acid transport protein 2, Fatty-acid-coenzyme A ligase, very long-chain 1, hFACVL1, HsT17226, Long-chain-fatty-acid--CoA ligase, SLC27A2, solute carrier family 27 (fatty acid transporter), member 2, Solute carrier family 27 member 2, THCA-CoA ligase, very-long-chain acyl-CoA synthetase, VLACS, VLACSVery long-chain-fatty-acid-CoA ligase, VLCS, VLCSvery long-chain acyl-CoA synthetase | |
| HEK293 human embryonic kidney cell line transfected with human FATP2 Met1-Leu620 Accession # O14975 | |
| RUO | |
| 11001 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 466106 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O14975 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FATP2. Stains human FATP2 transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction