missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FATP1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC3304R-100UG
This item is not returnable.
View return policy
Description
FATP1/SLC27A1 Monoclonal specifically detects FATP1/SLC27A1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| FATP1/SLC27A1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6PCB7 | |
| SLC27A1 | |
| NS0 mouse myeloma cell line transfected with human FATP1 Met1-Leu646 Accession # Q6PCB7 | |
| RUO | |
| 376497 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 308420 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ACSVL5FATP-1, EC 6.2.1, EC 6.2.1.-, FATP1Solute carrier family 27 member 1, FATPlong-chain fatty acid transport protein 1, Fatty acid transport protein 1, FLJ00336, MGC71751, SLC27A1, solute carrier family 27 (fatty acid transporter), member 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FATP1 in cell transfectants but not non-transfected parent cells in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction