missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Fascin ELISA Kit (Chemiluminescence)

Product Code. 18364931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364931 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364931 Supplier Novus Biologicals™ Supplier No. NBP275350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human Fascin ELISA Kit (Chemiluminescence) measures human Fascin in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16-10 ng/mL
Assay Sensitivity 0.09 ng/mL
Cross Reactivity This kit recognizes Human FSCN in samples. No significant cross-reactivity or interference between Human FSCN and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 6624
Gene Symbol FSCN1
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Fascin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.