missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fas/TNFRSF6/CD95 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF326
This item is not returnable.
View return policy
Description
Fas/TNFRSF6/CD95 Polyclonal specifically detects Fas/TNFRSF6/CD95 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Fas/TNFRSF6/CD95 | |
| Polyclonal | |
| Biotin | |
| Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 355 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| FAS | |
| Mouse myeloma cell line NS0-derived recombinant human Fas | |
| 50 μg | |
| Primary | |
| Detects human Fas in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant human Fas Ligand and recombinant mouse Fas is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction