missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fas Ligand/TNFSF6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
Brand: R&D Systems MAB126-SP
This item is not returnable.
View return policy
Description
Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Human samples. It is validated for Flow Cytometry, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Fas Ligand/TNFSF6 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 1-5 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P48023 | |
| FASLG | |
| Chinese hamster ovary cell line CHO-derived recombinant human Fas Ligand/TNFSF6 Pro134-Leu281 Accession # P48023 | |
| 25 μg | |
| Primary | |
| Detects human Fas Ligand/TNFSF6 in ELISAs. | |
| Human | |
| Purified |
| Neutralization, ELISA | |
| 100419 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 356 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction