missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human FAP QuicKit ELISA
Specifications
Specifications
| Assay Range | 54.70 to 3500 pg/mL |
| Assay Sensitivity | 9.01 pg/mL |
| Sample Type | Cell Culture Supernatant, Serum, Plasma (EDTA), Plasma (Heparin) |
| Specificity | Natural and recombinant human FAP. |
| Target Species | Human |
| Gene ID (Entrez) | 2191 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?