missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM96B (aa 18-91) Control Fragment Recombinant Protein

Product Code. 30182131
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182131

Brand: Invitrogen™ RP98714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59598 (PA5-59598. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Signs of Rubinstein-Taybi include mental retardation and predisposition to tumor growth and white blood cell neoplasias. Crohn's disease is a gastrointestinal inflammatory condition associated with chromosome 16 through the NOD2 gene. An association with systemic lupus erythematosis and a number of other autoimmune disorders with the pericentromeric region of chromosome 16 has led to the identification of SLC5A11 as a potential autoimmune modifier. The FAM96B gene product has been provisionally designated FAM96B pending further characterization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y3D0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51647
Name Human FAM96B (aa 18-91) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110019N10Rik; CGI-128; CIAB; Ciao2b; Cytosolic iron-sulfur assembly component 2 B; Fam96b; family with sequence similarity 96 member B; family with sequence similarity 96, member B; HSPC118; MIP18; Mitotic spindle-associated MMXD complex subunit MIP18; MSS19-interacting protein of 18 kDa; Protein FAM96B; RGD1311762
Common Name FAM96B
Gene Symbol CIAO2B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LIYQRSGERPVTAGEEDEQVPDSIDAREIFDLIRSINDPEHPLTLEELNVVEQVRVQVSDPESTVAVAFTPTIP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.