missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM65A (aa 849-965) Control Fragment Recombinant Protein

Product Code. 30206089
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206089

Brand: Invitrogen™ RP100912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52228 (PA5-52228. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Downstream effector protein for Rho-type small GTPases that plays a role in cell polarity and directional migration (PubMed:27807006). Acts as an adapter protein, linking active Rho proteins to STK24 and STK26 kinases, and hence positively regulates Golgi reorientation in polarized cell migration upon Rho activation (PubMed:27807006). Involved in the subcellular relocation of STK26 from the Golgi to cytoplasm punctae in a Rho- and PDCD10-dependent manner upon serum stimulation (PubMed:27807006). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6ZS17
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79567
Name Human FAM65A (aa 849-965) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310066E14Rik; AI837433; Fam65a; family with sequence similarity 65 member A; family with sequence similarity 65, member A; FLJ13725; Kiaa1930; protein FAM65A; Rho family-interacting cell polarization regulator 1; RIPOR1
Common Name FAM65A
Gene Symbol RIPOR1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HALESFSFLNEDEDEDNDVPGDRPPSSPEAGAEDSIDSPSARPLSTGCPALDAALVRHLYHCSRLLLKLGTFGPLRCQEAWALERLLREARVLEAVCEFSRRWEIPASSAQEVVQFS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.