missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FAM53C (aa 260-358) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Packungsgröße:
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60125 (PA5-60125. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene belongs to the FAM53 protein family. FAM53 protein family members bind to a transcriptional regulator that modulates cell proliferation. Alternative splicing results in multiple transcript variants.
Spezifikation
Spezifikation
| Accession Number | Q9NYF3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51307 |
| Name | Human FAM53C (aa 260-358) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2810012G03Rik; C5orf6; FAM53C; family with sequence similarity 53 member C; family with sequence similarity 53, member C; Gm21179; Protein FAM53C; putative nuclear protein |
| Common Name | FAM53C |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur