missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM40A (aa 323-382) Control Fragment Recombinant Protein

Product Code. 30211008
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211008

Brand: Invitrogen™ RP105154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66330 (PA5-66330. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the cortical actin filament dynamics and cell shape.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5VSL9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 85369
Name Human FAM40A (aa 323-382) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6330569M22Rik; 6530401O14Rik; Fam40a; family with sequence similarity 40, member A; FAR11 factor arrest 11 homolog A; FAR11A; homolog of yeast FAR11 protein 1; hypothetical protein; Kiaa1761; Protein FAM40A; QnpA-18494; RP4-773N10.1; striatin interacting protein 1; striatin-interacting protein 1; striatin-interacting protein 1; LOW QUALITY PROTEIN: striatin-interacting protein 1; Strip1
Common Name FAM40A
Gene Symbol STRIP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DSIKVIRNMRAASPPASASDLIEQQQKRGRREHKALIKQDNLDAFNERDPYKADDSREEE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.