missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3D Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2869
This item is not returnable.
View return policy
Description
FAM3D Monoclonal specifically detects FAM3D in Human samples. It is validated for Western Blot.
Specifications
| FAM3D | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q96BQ1 | |
| FAM3D | |
| Mouse myeloma cell line NS0-derived recombinant human FAM3D Trp39-Phe224 Accession # Q96BQ1 | |
| 100 μg | |
| Primary | |
| Detects human FAM3D in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) FAM3A, rhFAM3B, rhFAM3C, or recombinant mouse FAM3D is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 358203 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cytokine-like protein EF-7, EF7, family with sequence similarity 3, member D, OIT1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 131177 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction