missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3B Isoform B Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2866
This item is not returnable.
View return policy
Description
FAM3B Polyclonal specifically detects FAM3B in Human samples. It is validated for Western Blot.
Specifications
| FAM3B | |
| Polyclonal | |
| Unconjugated | |
| P58499 | |
| FAM3B | |
| Mouse myeloma cell line NS0-derived recombinant human FAM3B isoform B Glu30-Ser235 Accession # P58499 | |
| 100 μg | |
| Primary | |
| Detects human FAM3B in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant mouse FAM3B is observed and less than 5% cross-reactivity with recombinant human FAM3C is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| 2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PANDER, PRED44 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 54097 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction