missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM162A (aa 121-152) Control Fragment Recombinant Protein

Product Code. 30200465
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30200465 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30200465 Supplier Invitrogen™ Supplier No. RP105091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-66132 (PA5-66132, PA5-145111 (PA5-145111. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Proposed to be involved in regulation of apoptosis; the exact mechanism may differ between cell types/tissues. May be involved in hypoxia-induced cell death of transformed cells implicating cytochrome C release and caspase activation (such as CASP9) and inducing mitochondrial permeability transition. May be involved in hypoxia-induced cell death of neuronal cells probably by promoting release of AIFM1 from mitochondria to cytoplasm and its translocation to the nucleus; however, the involvement of caspases has been reported conflictingly.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96A26
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26355
Name Human FAM162A (aa 121-152) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310056P07Rik; C3orf28; DC16; E2IG5; E2-induced gene 5 protein; E2-induced gene 5 protein homolog; f162; Fam162a; family with sequence similarity 162 member A; family with sequence similarity 162, member A; FWP001; Growth and transformation-dependent protein; HGTD-P; HIF-1 alpha-responsive proapoptotic molecule; protein FAM162A
Common Name FAM162A
Gene Symbol FAM162A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EGKKAAQRHETLTSLNLEKKARLKEEAAMKAK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.