missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FAM129C (aa 133-210) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60306 (PA5-60306. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q86XR2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 199786 |
| Name | Human FAM129C (aa 133-210) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | B-cell novel protein 1; BCNP1; BCNP1 homolog; FAM129C; family with sequence similarity 129 member C; family with sequence similarity 129, member C; NIBAN3; niban-like protein 2; Protein FAM129C; Protein Niban 3 |
| Common Name | FAM129C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction