missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FABP5 (E-FABP) Recombinant Protein

Product Code. 11578881
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578881 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11578881 Supplier Invitrogen™ Supplier No. RP75555

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human epidermal FABP protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

FABP5 (fatty acid binding protein 5 (psoriasis-associated)) is a proteinthat in humans is encoded by theFABP5 gene, also known as PAFABP, EFABP, E-FABP, KFABP, PA-FABP, Epidermal-type fatty acid-binding protein, Fatty acid-binding protein 5, Psoriasis-associated fatty acid-binding protein homolog. It is a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. The PAFABP cDNA encodes a 135-amino acid protein with molecular weight 15,164. This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated inpsoriasistissue. It is though that FABPs roles include fatty acid uptake, transport, and metabolism. Kaczocha et al. (2009) identifiedFABP5and FABP7 as cytosolic proteins that transport AEA from the plasma membrane to subcellular fatty acid amide hydrolase, where it is hydrolyzed and inactivated. FABP3 did not show this specific transport function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q01469
Concentration 1.25 mg/mL
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 2171
Name Human FABP5 (E-FABP)
pH Range 8
Quantity 100 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias C-FABP; Cutaneous fatty acid-binding protein; DA11; EFABP; E-FABP; E-FABPPAFABP; Epidermal-type fatty acid-binding protein; epithelial fatty acid-binding protein; Fabp5; Fabpe; fatty acid binding protein 5; fatty acid binding protein 5 (psoriasis-associated); fatty acid binding protein 5, epidermal; fatty acid-binding protein 5; Fatty acid-binding protein, epidermal; fatty acid-binding protein, epidermal; uncharacterized protein LOC16592; keratinocyte lipid-binding protein; KFABP; Klbp; Mal1; PAFABP; PA-FABP; PA-FABPepidermal; Psoriasis-associated fatty acid-binding protein homolog
Common Name FABP5
Gene Symbol FABP5
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1- Glu135 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.