missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FABP5 (E-FABP) Recombinant Protein

Product Code. 11568881
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11568881 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11568881 Supplier Invitrogen™ Supplier No. RP75554

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Epidermal FABP (human) Recombinant Protein, RP-75554, from Invitrogen.

This product contains purified recombinant human epidermal FABP protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q01469
For Use With (Application) Control, Western Blot
Formulation 20mM tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 2171
Name Human FABP5
pH Range 8
Quantity 10 μg
Source E. coli
Immunogen N/A
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Fatty Acid Binding Protein
Common Name Human FABP5
Gene Symbol FABP5
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.