missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human FABP4/A-FABP DuoSet ELISA

Product Code. 18786577
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18786577 1 Kit (for 5 Plates) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18786577 Supplier R&D Systems™ Supplier No. DY315005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs

FABP4 (Fatty Acid Binding Protein 4), also known as A-FABP and aP2, is the predominant FABP found in adipocytes and is often used as a marker for adipocyte differentiation. It is also expressed in macrophages, dendritic cells, and endothelial cells. FABP4 is a key mediator of intracellular fatty acid transport and metabolism in adipose tissue. Its expression is regulated by multiple factors including fatty acids, PPAR gamma agonists, and Insulin, and its levels increase with lipolytic stimulation. It can increase the hydrolytic activity of Hormone-Sensitive Lipase and increase the production of proinflammatory cytokines and chemokines. FABP4 upregulation in adipocytes and macrophages is associated with the development of Insulin resistance, hypertriacylglycerolaemia, and atherosclerosis. Serum levels of FABP4 are elevated in obesity and metabolic syndrome, type 2 diabetes, HIV-associated lipodystrophy, polycystic ovary syndrome, nonalcoholic fatty liver disease, atherosclerosis, and acute ischaemic stroke. FABP4 is also overexpressed in multiple cancer types including ovarian, bladder, glioblastoma, and oral, and it may play a role in tumor progression.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit (for 5 Plates)
Species Human
Binding Target FABP4/A-FABP
For Use With (Application) ELISA
Gene ID (Entrez) 2167
Synonym Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, AFABP, A-FABP, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.