missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FABP2 (I-FABP) Recombinant Protein
Description
This product contains purified recombinant human intestinal FABP protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
May play roles in fatty acid transport and compartmentalization.
Specifications
Specifications
| Accession Number | P12104 |
| Concentration | 0.74 |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 2169 |
| Name | Human FABP2 (I-FABP) |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?