missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FABP1/L-FABP Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £361.00
Specifications
| Antigen | FABP1/L-FABP |
|---|---|
| Clone | 965616 |
| Dilution | Western Blot 2μg/mL, Immunocytochemistry 2-25μg/mL |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18141581
![]() |
R&D Systems
MAB29644-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18196031
![]() |
R&D Systems
MAB29644-100 |
100 μg |
£361.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP1/L-FABP Monoclonal specifically detects FABP1/L-FABP in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| FABP1/L-FABP | |
| Western Blot 2μg/mL, Immunocytochemistry 2-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P07148 | |
| 2168 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 965616 | |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, LFABP, L-FABP, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
| E. coli-derived recombinant human FABP1/L-FABP Met1-Ile127 Accession # P07148 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human FABP-1/L-FABP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title