missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EXOSC4 (aa 90-162) Control Fragment Recombinant Protein

Product Code. 30209337
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209337

Brand: Invitrogen™ RP109079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Non-catalytic component of the RNA exosome complex which has 3'-5' exoribonuclease activity and participates in a multitude of cellular RNA processing and degradation events. In the nucleus, the RNA exosome complex is involved in proper maturation of stable RNA species such as rRNA, snRNA and snoRNA, in the elimination of RNA processing by-products and non-coding pervasive transcripts, such as antisense RNA species and promoter-upstream transcripts (PROMPTs), and of mRNAs with processing defects, thereby limiting or excluding their export to the cytoplasm. The RNA exosome may be involved in Ig class switch recombination (CSR) and/or Ig variable region somatic hypermutation (SHM) by targeting AICDA deamination activity to transcribed dsDNA substrates. In the cytoplasm, the RNA exosome complex is involved in general mRNA turnover and specifically degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3' untranslated regions, and in RNA surveillance pathways, preventing translation of aberrant mRNAs. It seems to be involved in degradation of histone mRNA. The catalytic inactive RNA exosome core complex of 9 subunits (Exo-9) is proposed to play a pivotal role in the binding and presentation of RNA for ribonucleolysis, and to serve as a scaffold for the association with catalytic subunits and accessory proteins or complexes. EXOSC4 binds to ARE-containing RNAs.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NPD3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54512
Name Human EXOSC4 (aa 90-162) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110039I09Rik; 1500001N04Rik; EXOSC4; exosc4 protein; exosome complex component RRP41; exosome complex exonuclease RRP41; exosome complex exonuclease RRP41; exosome complex component RRP41; exosome component 4; exosome component Rrp41; FLJ20591; hRrp41p; p12A; ribosomal RNA processing protein 41; ribosomal RNA-processing protein 41; Rrp41; RRP41A; Rrp41p; SKI6; Ski6p; Unknown (protein for MGC:134035); zgc:73175
Common Name EXOSC4
Gene Symbol EXOSC4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PHGDRKSCEMGLQLRQTFEAAILTQLHPRSQIDIYVQVLQADGGTYAACVNAATLAVLDAGIPMRDFVCACSA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.