missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EXOC6 (aa 227-267) Control Fragment Recombinant Protein

Product Code. 30206576
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206576

Brand: Invitrogen™ RP97094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57595 (PA5-57595. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The product of this gene belongs to the SEC15 family. It is highly similar to the protein encoded by Saccharomyces cerevisiae SEC15 gene. This protein is essential for vesicular traffic from the Golgi apparatus to the cell surface in yeast. It is one of the components of a multiprotein complex required for exocytosis. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TAG9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54536
Name Human EXOC6 (aa 227-267) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4833405E05Rik; AW413330; C430002C19; DKFZp761I2124; Exoc6; EXOC6A; Exocyst complex component 6; exocyst complex component Sec15A; FLJ1125; FLJ11251; hbd; MGC33397; msec15; rSec15; rsec15-like protein; Sec15; SEC15 homolog; SEC15A; SEC15L; SEC15L1; SEC15L3; SEC15-like 1; SEC15-like protein 1; SEC15-like protein 3; Sec15p; zgc:77548
Common Name EXOC6
Gene Symbol Exoc6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TFSVSLQKQNKMKFGKNMYINRDRIPEERNETVLKHSLEEE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado