missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ETV5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7107
This item is not returnable.
View return policy
Description
ETV5/ERM Monoclonal specifically detects ETV5/ERM in Human samples. It is validated for Immunocytochemistry.
Specifications
| ETV5/ERM | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P41161 | |
| ETV5 | |
| E. coli-derived recombinant human ETV5 Asn119-Ser223 Accession # P41161 | |
| 100 μg | |
| Primary | |
| Detects human ETV5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ETV1 or rhETV2 is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 654404 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ERM, ERMEts-related protein ERM, ETS translocation variant 5, ets variant 5, ets variant gene 5 (ets-related molecule), ets-related molecule | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2119 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction