missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ESPN (aa 785-848) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55941 (PA5-55941. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Multifunctional actin-bundling protein. Plays a major role in regulating the organization, dimensions, dynamics and signaling capacities of the actin filament-rich, microvillus-type specializations that mediate sensory transduction in variouS mechanosensory and chemosensory cells.
Specifications
Specifications
| Accession Number | B1AK53 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 83715 |
| Name | Human ESPN (aa 785-848) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | actin cytoskeletal regulatory protein; autosomal recessive deafness type 36 protein; DFNB36; Ectoplasmic specialization protein; espin; Espn; je; jerker; Jerker, deafness locus; LP2654 |
| Common Name | ESPN |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction