missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin R Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3072
This item is not returnable.
View return policy
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot.
Specifications
| Erythropoietin R | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P19235 | |
| EPOR | |
| A synthetic phosphopeptide corresponding to residues surrounding Y426 of human Erythropoietin R Accession # P19235 | |
| 100 μg | |
| Primary | |
| Detects human Erythropoietin R in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 690748 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2057 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction