missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, Immunoaffinity Purification.
Specifications
Specifications
| Antigen | Erythropoietin/EPO |
| Applications | Western Blot, Affinity Purification |
| Classification | Monoclonal |
| Clone | AE7A5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunoaffinity Purification |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P01588 |
| Gene Alias | ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 |
| Gene Symbols | EPO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?