missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ERK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1940-SP
This item is not returnable.
View return policy
Description
ERK1 Monoclonal specifically detects ERK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| ERK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 0.3-25 ug/mL | |
| P27361 | |
| MAPK3 | |
| Human ERK1 synthetic peptide Met1-Val28 Accession # P27361 | |
| 25 μg | |
| Primary | |
| Detects human ERK1 in Western blots. In Western blots, no cross-reactivity with human ERK2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 250603 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MAPK3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, P44MAPK, PRKM3, PRKM3EC 2.7.11.24 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5595 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction