missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Erbin (aa 582-662) Control Fragment Recombinant Protein

Product Code. 30212774
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212774

Brand: Invitrogen™ RP105141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66288 (PA5-66288. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96RT1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55914
Name Human Erbin (aa 582-662) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700028E05Rik; densin-180-like protein; epididymis secretory protein Li 78; Erbb2 interacting protein; ERBB2 interacting protein-like; erbb2-interacting protein; Erbb2ip; Erbin; HEL-S-78; Kiaa1225; Lap2; mKIAA1225; Protein LAP2; RGD1562952; similar to Erbb2 interacting protein isoform 2
Common Name Erbin
Gene Symbol ERBIN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SENLKHIVNHDDVFEESEELSSDEEMKMAEMRPPLIETSINQPKVVALSNNKKDDTKETDSLSDEVTHNSNQNNSNCSSPS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.