missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 23-652 of Human ErbB2/Her2 The Recombinant Human ErbB2/Her2 Protein is derived from Baculovirus. The Recombinant Human ErbB2/Her2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | In PBS (pH 7.4) containing 10% glycerol |
| Molecular Weight (g/mol) | 70.4 kDa |
| Name | ERBB2 Recombinant Protein |
| Quantity | 10 μg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1.0 EU per 1μg of protein (determined by LAL method) |
| Gene Alias | CD340 antigen, CD340 |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?