missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human ErbB2/Her2 Recombinant Protein

Product Code. 18210444 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18210444

Brand: Novus Biologicals™ NBP25965350UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 23-652 of Human ErbB2/Her2 The Recombinant Human ErbB2/Her2 Protein is derived from Baculovirus. The Recombinant Human ErbB2/Her2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation In PBS (pH 7.4) containing 10% glycerol
Molecular Weight (g/mol) 70.4 kDa
Name ERBB2 Recombinant Protein Antigen
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias CD340 antigen, CD340
Gene Symbol ERBB2
Product Type Recombinant Protein Antigen
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade >85% by SDS-PAGE
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.