missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EQTN (aa 35-152) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (51%), Rat (51%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53312 (PA5-53312. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q9NQ60 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 54586 |
| Name | Human EQTN (aa 35-152) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | acrosome formation associated factor; acrosome formation-associated factor; AFAF; C9orf11; EQTN; Equatorin; equatorin, sperm acrosome associated; SPACA8; sperm acrosome associated 8 |
| Common Name | EQTN |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction