missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human EPO (Erythropoietin) Recombinant Protein
Erythropoietin (EPO) is a bioactive protein intended for use in cell culture applications
£344.00 - £1658.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 18.4 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | EPO (Erythropoietin) |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12313703
|
Gibco™
PHC9634 |
10μg |
£344.00
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12303703
|
Gibco™
PHC9631 |
100μg |
£1658.00
Each |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human EPO (Erythropoietin) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Erythropoietin is a member of the EPO/TPO family. It regulates red cell production by promoting erythroid differentiation and initiating hemoglobin synthesis. This protein also has neuroprotective activity against a variety of potential brain injuries and antiapoptotic functions in several tissue types.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by in vivo bioassay measuring dose-dependent increase in reticulocyte count with normocythaemic mice
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Approved for shipment on Wet or Dry Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Proliferation of TF1 cells | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Stem Cell Research, Neurobiology | |
95 % |
18.4 kDa | |
EPO (Erythropoietin) | |
Store at 2 to 8°C | |
0.8 ng/ml | |
< 0.1 ng/μg | |
Mammalian | |
Recombinant, Ligand | |
EPO | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.