missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Epiregulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1425-SP
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for Western Blot.
Specifications
| Epiregulin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O14944 | |
| EREG | |
| E. coli-derived recombinant human Epiregulin Val63-Leu108 Accession # O14944 | |
| 25 μg | |
| Primary | |
| Detects the EGF-like domain of human Epiregulin in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 100% cross-reactivity with recombinant mouse Epiregulin. | |
| Human | |
| Purified |
| Western Blot | |
| 183625 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| epiregulin, ER, EREG, proepiregulin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2069 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction