missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Epiregulin Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC14251N-100UG
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Epiregulin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O14944 | |
| EREG | |
| E. coli-derived recombinant human Epiregulin Val63-Leu108 Accession # O14944 | |
| RUO | |
| 2069 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 183629 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| epiregulin, ER, EREG, proepiregulin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Epiregulin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Betacellulin, rhEGF, recombinant mouse Epiregulin, or rhHB-EGF is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction