missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ephrin-B3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB395
This item is not returnable.
View return policy
Description
Ephrin-B3 Monoclonal specifically detects Ephrin-B3 in Human samples. It is validated for Western Blot.
Specifications
| Ephrin-B3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EFL-6, Efnb3, Elk-L3, EPH-related receptor tyrosine kinase ligand 8, Ephrin B3, ephrin-B3, EphrinB3, Epl8, EPLG8EPH-related receptor transmembrane ligand ELK-L3, LERK-8, LERK8EFL6, NLERK-2 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 1949 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 88838 | |
| Western Blot 1 ug/mL | |
| Q15768 | |
| EFNB3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Ephrin-B3 Leu28-Ser224 Accession # Q15768 | |
| 500 μg | |
| Primary | |
| Detects human Ephrin-B3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Ephrin-A1, rmEphrin-A2, recombinant human (rh) Ephrin-A3, rhEphrin-A4, rhEphrin-A5, rmEphrin-B1, and rmEphrin-B2 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction