missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EphB3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5667
This item is not returnable.
View return policy
Description
EphB3 Monoclonal specifically detects EphB3 in Human samples. It is validated for Western Blot.
Specifications
| EphB3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P54753 | |
| EPHB3 | |
| Mouse myeloma cell line NS0-derived recombinant human EphB3 Leu38-Ala550 Accession # P54753 | |
| 100 μg | |
| Primary | |
| Detects human EphB3 in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant mouse (rm) EphB3, 25% cross-reactivity with rmEphB2, and no cross-reactivity with recombinant human (rh) EphA1, A2, A5, A6, A10, rmEphA3, A4, A7, B4, B6, or recombinant rat EphB1. | |
| Human | |
| Purified |
| Western Blot | |
| 647308 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cek10, EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Mdk5, Sek4, Tyro6, Tyrosine-protein kinase TYRO6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2049 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction