missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EphB3 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB56671R-100UG
This item is not returnable.
View return policy
Description
EphB3 Monoclonal specifically detects EphB3 in Human samples. It is validated for Flow Cytometry.
Specifications
| EphB3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P54753 | |
| EPHB3 | |
| Mouse myeloma cell line NS0-derived recombinant human EphB3 Leu38-Ala550 Accession # P54753 | |
| RUO | |
| 2049 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 647354 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Cek10, EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Mdk5, Sek4, Tyro6, Tyrosine-protein kinase TYRO6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human EphB3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) EphA3, rhEphA4, EphB2, or recombinant mouse EphB3 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering