missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EphA2 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF3035
This item is not returnable.
View return policy
Description
EphA2 Polyclonal specifically detects EphA2 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| EphA2 | |
| Polyclonal | |
| Biotin | |
| P29317 | |
| EPHA2 | |
| Mouse myeloma cell line NS0-derived recombinant human EphA2 Gln25-Asn534 Accession # P29317 | |
| 50 μg | |
| Primary | |
| Detects human EphA2 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse (rm) EphA2 is observed and less than 1% cross-reactivity with recombinant human (rh) EphA1, rmEphA4, rhEphA5, rmEphA6, rmEphA7, and rmEphA8 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
| ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1969 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction