missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EphA10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £399.00
Specifications
| Antigen | EphA10 |
|---|---|
| Clone | 494130 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754678
![]() |
R&D Systems
MAB5188-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18101831
![]() |
R&D Systems
MAB5188 |
100 μg |
£399.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EphA10 Monoclonal specifically detects EphA10 in Human samples. It is validated for Immunohistochemistry.Specifications
| EphA10 | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q5JZY3 | |
| 284656 | |
| E. coli-derived recombinant human EphA10 Glu34-Glu283 (predicted) Accession # Q5JZY3 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human EphA10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human EphA1, A2, A5, A6, B2, B3, B4, B6, recombinant mouse EphA3, A4, or A8 is observed. |
| 494130 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10.1, EPH receptor A10, EphA10s protein, ephrin type-A receptor 10, FLJ16103, FLJ33655, MGC43817 | |
| EPHA10 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title