missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Enteropeptidase/Enterokinase Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1585
This item is not returnable.
View return policy
Description
Enteropeptidase/Enterokinase Polyclonal specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Enteropeptidase/Enterokinase | |
| Polyclonal | |
| Unconjugated | |
| P98073 | |
| TMPRSS15 | |
| Mouse myeloma cell line NS0-derived recombinant human Enteropeptidase/Enterokinase Leu41-His1019 Accession # P98073 | |
| 100 μg | |
| Primary | |
| Detects human Enteropeptidase/Enterokinase in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTK, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7, PRSS7enteropeptidase, Serine protease 7, TMPRSS15, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5651 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction