missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Enteropeptidase/Enterokinase Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1585
This item is not returnable.
View return policy
Description
Enteropeptidase/Enterokinase Monoclonal specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Enteropeptidase/Enterokinase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| NP_002763 | |
| TMPRSS15 | |
| Mouse myeloma cell line NS0-derived recombinant human Enteropeptidase/Enterokinase Leu41-His1019 Accession # NP_002763 | |
| 500 μg | |
| Primary | |
| Detects human Enteropeptidase/Enterokinase in direct ELISAs and Western blots. It recognizes all EK forms containing the heavy chain including the single chain precursor and the mature heterodimer consisting of a heavy and light chain. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Factor X and rhThrombin is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 190034 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTK, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7, PRSS7enteropeptidase, Serine protease 7, TMPRSS15, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5651 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction