missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ENPP-7/Alk-SMase Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4924-SP
This item is not returnable.
View return policy
Description
ENPP-7/Alk-SMase Monoclonal specifically detects ENPP-7/Alk-SMase in Human samples. It is validated for Western Blot.
Specifications
| ENPP-7/Alk-SMase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q6UWV6 | |
| ENPP7 | |
| Mouse myeloma cell line NS0-derived recombinant human ENPP-7/Alk-SMase Pro23-Ser439 Accession # Q6UWV6 | |
| 25 μg | |
| Primary | |
| Detects human ENPP-7/Alk-SMase in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ENPP-2, rhENPP-5, recombinant mouse ENPP-7 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 560514 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, ENPP7, Intestinal alkaline sphingomyelinase, MGC50179, NPP7, NPP-7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 339221 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction