missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ENPP-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6136-SP
This item is not returnable.
View return policy
Description
ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Western Blot.
Specifications
| ENPP-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, ENPP1, Ly-41 Antigen, M6S1, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPP1, NPPS, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1, PCA1ARHR2, PDNP1, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5167 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q13822 (ENPP-2) P22413 (ENPP-1) | |
| ENPP1 | |
| Mouse myeloma cell line NS0-derived recombinant human ENPP-2-1-2 Asp49-Trp144 (ENPP-2), Val191-Leu591 (ENPP-1), Asn532-Ile863 (ENPP-2). Cross-reactivity with human ENPP-2 was removed from the final product. Accession # Q13822 (ENPP-2) P22413 (ENPP-1) | |
| 25 μg | |
| Primary | |
| Detects human ENPP-1 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human ENPP-2 and recombinant mouse ENPP-2 is observed after removal of cross-reactivity with human ENPP-2. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction