missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ENOX1 (aa 465-559) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58355 (PA5-58355. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Probably acts as a terminal oxidase of plasma electron transport from cytosolic NAD(P)H via hydroquinones to acceptors at the cell surface. Hydroquinone oxidase activity alternates with a protein disulfide-thiol interchange/oxidoreductase activity which may control physical membrane displacements associated with vesicle budding or cell enlargement. The activities oscillate with a period length of 24 minutes and play a role in control of the ultradian cellular biological clock.
Specifications
Specifications
| Accession Number | Q8TC92 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55068 |
| Name | Human ENOX1 (aa 465-559) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | bA64J21.1; candidate growth-related and time keeping constitutive hydroquinone (NADH) oxidase; candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase; cCNOX; Cell proliferation-inducing gene 38 protein; CNOX; Constitutive Ecto-NOX; ecto-NOX disulfide-thiol exchanger 1; ENO x 1; Hydroquinone [NADH] oxidase; PIG38; Protein disulfide-thiol oxidoreductase |
| Common Name | ENOX1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction