missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human eNOS/NOS3 ELISA Kit

Product Code. 17164919
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17164919 5 x 96 Tests Each
17854080 10 x 96 Tests Pack of 10
17114453 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17164919 Supplier Invitrogen™ Supplier No. EH169RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human eNOS/NOS3 quantitates human eNOS/NOS3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human eNOS/NOS3.

Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) and endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2310065A03Rik; cNOS; constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNOS; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183
Assay Range 0.4 to 100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4846
Gene Symbol NOS3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target eNOS
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.