missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human eNOS Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF950-SP
This item is not returnable.
View return policy
Description
eNOS Polyclonal specifically detects eNOS in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.
Specifications
| eNOS | |
| Polyclonal | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 0.3 ug/10^6 cells | |
| Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOS3, NOSIII | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4846 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NOS3 | |
| E. coli-derived recombinant human eNOS | |
| 25 μg | |
| Primary | |
| Detects human eNOS in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction