missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endothelin-3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5579
This item is not returnable.
View return policy
Description
Endothelin-3 Monoclonal specifically detects Endothelin-3 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Endothelin-3 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P14138 | |
| EDN3 | |
| Human Endothelin-3 synthetic peptide Accession # P14138 | |
| 100 μg | |
| Primary | |
| Detects human Endothelin-3. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 3B6 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EDN3, endothelin 3, endothelin-3, Endothelin3, ET3, ET-3, HSCR4, MGC15067, MGC61498, PPET3, preproendothelin-3, truncated endothelin 3, WS4B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1908 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction