missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endoglin/CD105 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1097-SP
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot.
Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q5T9B9 | |
| ENG | |
| Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105 Glu26-Gly256 Accession # Q5T9B9 | |
| 25 μg | |
| Primary | |
| Detects human Endoglin/CD105 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 10-50% cross-reactivity with recombinant mouse Endoglin. | |
| Human | |
| Purified |
| Western Blot | |
| 166709 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2022 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction